Learn More
PEG10 Antibody (981902) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB99751J
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PEG10 Monoclonal antibody specifically detects PEG10 in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOF
Spezifikation
| PEG10 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23089 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Flow Cytometry (Intracellular Staining), CyTOF | |
| 981902 | |
| ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, Mart2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3MEF3-like protein 1, Ty3/Gypsy-like protein | |
| Synthetic peptide containing human PEG10, Accession # Q86TG7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |