missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12323-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Spezifikation
PAX6 | |
Polyclonal | |
Unconjugated | |
PAX6 | |
AN, AN2, Aniridia type II protein, D11S812E, MGDA, Oculorhombin, paired box 6, Paired box protein Pax 6, PAX6, WAGR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18508, 5080 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26367, P63015 | |
PAX6 | |
PAX6 Fusion Protein Ag2984 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |