missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Pro Star PD216 pH/ion and DO Bench Meter

Artikelnummer. 30245795
Click to view available options
Produkttyp:
Messgerät mit Fuß
Messgerät mit Standardkit
Messgerät mit pH-/BSB-Kit
Messgerätt mit pH-/optischem DO-Kit
Packungsgröße:
1 Set
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30245795

Marke: Thermo Scientific™ PSTAR2165

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Designed to provide accurate and reliable pH, ORP, ion concentration, dissolved oxygen, barometric pressure and temperature analysis, the Thermo Scientific™ Orion™ Pro Star PD216 bench meter is well suited for laboratories that desire a modern meter with advance performance.

Designed to provide accurate and reliable pH, ORP, ion concentration, DO, barometric pressure and temperature analysis, the Orion Pro Star PD216 bench meter is well suited for laboratories that desire a modern meter with advance performance. Featuring a 7-in., high-resolution color display with responsive capacitive touchscreen, the Orion Pro Star meter focuses on ensuring an easy to use, innovative meter for your testing needs. The Orion Pro Star meter offers onscreen guidance with step-by-step instructions and supports GLP compliance with date and time stamped calibration and data logs.

Orion Pro Star PD215 pH/ion and dissolved oxygen bench meters are designed to provide accurate and reliable testing in laboratories that need a modern, easy-to-operate meter with advanced performance and reliable data reporting. Orion Pro Star bench meter benefits include:

  • Large 7-in., high-resolution color display
  • Responsive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps optimize bench space
  • Supports GLP compliance
  • Stores 20 calibration logs per parameter with date and time
  • Stores 10,000 data log sets with date and time
  • Outputs to a printer, computer, or USB flash drive for long-term data retention
  • Inputs sample ID with optional USB barcode scanner, USB keyboard or USB mouse
  • Stirs samples with meter-controlled and meter-powered stirrer probes

Orion Pro Star PD215 pH/ion and DO meters utilize two channels to offer the following measurements:

  • pH, mV, ORP or ion concentration with temperature
  • Dissolved oxygen as % saturation or concentration in ppm or mg/L with temperature

First time meter startup is made easy with the meter setup wizard, which guides the user through important meter and measurement settings. Utilize onscreen meter tours and guides for calibration, measurements, meter connections, electrode stand assembly and pH electrode usage to better understand meter operation, warnings and required maintenance. Use guided calibrations with step-by-step instructions to perform calibrations correctly and ensure accurate, reliable and reportable measurements.

Easily view measurements on the large 7-in. color graphic touchscreen display. View measurement channels individually or two simultaneously to meet changing measurement needs. Select the basic, advanced or custom measure display format to view only the information you prefer in the measure mode. Select the graph view to display individual measurements in a graph with temperature for measurement versus time studies.

While taking measurements, easily enter sample identification information into the meter using an optional meter accessory including USB barcode scanner, USB keyboard or USB mouse. Easily and uniformly stir samples using an optional meter-controlled and meter-powered stirrer probe, which eliminates the need for a magnetic stir plate and bars.

Save, display and export up to 20 calibrations per parameter with date and time stamp. Filter calibration logs by user and export calibration logs by date range or all at once.

Easily save measurements to the data log for long-term data retention. Save, display and export up to 10,000 data log sets with date and time stamp. View individual data log sets with extensive measurement information organized into an easy-to-read onscreen list format. Filter data logs by user, date and time or log number and export data logs by date and time range, log number range or all at once.

Output meter data logs, calibration logs and methods to a printer, computer or USB flash drive. Save data to a computer or USB drive as comma separated value (CSV) files for import into programs like Microsoft Excel or save data to a USB drive as report files for printable records. Customize exported files to include all data, advanced, intermediate or basic data only to obtain only the information needed for your record keeping. Utilize bidirectional communication with remote commands to interface the meter with computer programs including LIMS, HyperTerminal, PuTTY and additional data collection software.

Save measurement settings to standardize your analysis and SOPs using meter methods. Save up to 10 methods per parameter. Import methods to and export methods from other Orion Pro Star series meters to standardize your testing across multiple meters. Also save, import and export incremental techniques for determination of ion concentration to standardize your known addition, known subtraction, analyte addition and analyte subtraction analyses.

Activate the secure mode and assign up to 3 administrators and up to 30 users with individual account logins and password protection. Administrators can select from pre-defined user access levels to meter functions. Control access to date and time settings, instrument settings, data logs, and calibration logs, or customize each user's access level from full access to basic data collection only. Administrators can also export the user list and audit trail to help ensure digital data integrity.

Operate the included electrode stand to facilitate secure, smooth movement of electrodes into and out of solutions. The electrode stand's weighted base provides flexible placement of the stand around the meter. Secure a wide range of electrodes using the various electrode holder options. Neatly organize electrode cables in the built-in side channels of the stand.

TRUSTED_SUSTAINABILITY

Spezifikation

Netzteil Universal 100-240 V 50/60 Hz power supply with wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Ausgänge USB flash drive, computer or printer (STARA-106)
Genauigkeit (Barometer) ±6.0 mmHg; ±0.008 bar
Genauigkeit (DO) ±2% saturation; ±0.02 mg/L or ppm
Genauigkeit (ISE) ±0.2 mV or ±0.05% of the reading, whichever is greater
Genauigkeit (mV) ±0.2 mV or ±0.05% of the reading, whichever is greater
Genauigkeit (ORP) ±0.2 mV or ±0.05% of the reading, whichever is greater
Genauigkeit (pH) ±0.002 pH
Genauigkeit (DO, polarographisch) ±2% saturation; ±0.02 mg/L or ppm
Genauigkeit (RDO) ±2% saturation; ±0.02 mg/L or ppm
Erweiterte ISE-Funktionen Single known addition, single known subtraction, double known addition, double known subtraction, analyte addition and analyte subtraction
Alarmausgang Calibration reminder alarm, high and low limit alarm per parameter
Hintergrundbeleuchtungsoption Yes
Luftdruckkorrektur: Yes, automatic with built-in barometer or manual
Kalibrierung pH, ORP, Ion Concentration, DO, Barometric Pressure, Temperature
Kalibrierungsmodi Water-saturated air (100% and 0% saturation), air-saturated water (100% and 0% saturation) or custom concentration value (Winkler)
Kalibrierpunkte 1 to 5 (pH), 1 (ORP), 1 to 5 (ion), 1 to 2 (DO), 1 (barometer), 1 to 2 (temperature)
Kalibrierpunkte (ISE) Up to 5
Kalibrierpunkte (pH) 1 to 5 points
Zertifizierung/Konformität CE, TUV 3-1, FCC Class A
Datenpunkte 10,000 with Date and Time Stamp
Kanäle 2
Displaytyp 7-in. High-resolution Color Graphic LCD
IP-Bewertung IP54
Measurement Modes pH, ORP, mV, Ion Concentration (ISE), Dissolved Oxygen (mg/L), Dissolved Oxygen (% Saturation), Temperature, Barometric Pressure
Percent Saturation Range Polarographic DO 0.0 to 600.0% saturation
Bereich (mV) -2000 to 2000 mV
Bereich (pH) -2 to 20
Auflösung (ISE) 0.0001 minimum, 1 to 5 digits
Auflösung (pH) 0.1, 0.01 or 0.001 pH
Temperaturbereich (metrisch) -30.0°C to +130.0°C
Zur Verwendung mit (Anwendung) Most common pH samples as well as pH samples containing tris buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; wide variety of dissolved oxygen samples using polarographic DO electrode
Enthält Meter with stand (PSTAR2160); ROSS Ultra Triode refillable epoxy-body pH/ATC electrode (8157BNUMD); polarographic DO/ATC electrode with calibration sleeve (083005MD); pH 4, 7, 10 buffers and rinse solution pouches; pH electrode storage solution and cleaning solution; pH electrode storage sleeve and base; polarographic DO electrode maintenance kit; BOD adapter
Eingänge Channel 1: BNC for pH, ORP or ISE, 8-pin MiniDIN for ATC, pin-tip for reference; Channel 2: 9-pin MiniDIN for dissolved oxygen with ATC
Isopotentialpunkt 7.000 pH; 1.000 ion concentration
Tastenfeld Capacitive touchscreen user interface
Sprachen Chinese, English, French, German, Italian, Japanese, Korean, Portuguese, Spanish
Art der Protokollfunktion Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Hauptspeicher Non-volatile
Verfahren 10 per channel (20 total)
Modell PD216 Bench Meter Standard pH and DO Kit
Percent Saturation Relative Accuracy (RDO) ±2% saturation
Percent Saturation Resolution (Polarographic DO) 0.1, 1% saturation
Sättigungsbereich (RDO) in % 0.0 to 600.0% saturation
Percent Saturation Relative Accuracy (Polarographic DO) ±2% saturation
Auflösung der Sättigung in % (RDO) 0.1, 1% saturation
Druckbereich 400.0 to 850.0 mmHg; 0.533 to 1.133 bar
Sondentyp Polarographic or optical DO electrode
Produkttyp Messgerät mit Standardkit
Bereich (DO) 0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Bereich (Ionen) 0.0001 to 19999
Bereich (ISE) 0.0001 to 19999
Bereich (ORP) -2000 to 2000 mV
Bereich (DO, polarographisch) 0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Bereich (RDO) 0.0 to 600.0% saturation; 0.01 to 90.0 mg/L or ppm
Bereich (relative mV) -2000 to 2000 mV
Auflösung (DO) 0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Auflösung (Ionen) 0.0001 minimum, 1 to 5 digits
Auflösung (mV) 0.1 mV
Auflösung (ORP) 0.1 mV
Auflösung (DO, polarographisch) 0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Auflösung (RDO) 0.1, 1% saturation; 0.01, 0.1 mg/L or ppm
Salinitätsfaktor 0.0 to 50.0 ppt
Temperaturgenauigkeit ±0.3°C, ±0.5°F
Temperaturfühlerkalibrierung 1 or 2 point temperature offset calibration
Temperaturbereich (angloamerikanisch) -22.0°F to +266.0°F
Temperaturauflösung 0.1°C, 0.1°F
Temperaturwahl Automatic with ATC temperature sensor, ambient or manual
Gewährleistung 2 year meter warranty
Abmessungen (L x B x H) 218.9 x 153.2 x 91.33 mm; 8.62 x 6.03 x 3.60 in.
Gewicht (angloamerikanisch) 2.20 lbs.
Gewicht (metrisch) 1.00 kg
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt