missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NU 9056, Tocris Bioscience™
Click to view available options
Menge:
10 mg
Beschreibung
Selective KAT5 (Tip60) histone acetyltransferase inhibitor (IC50 values are < 2, 60, 36, and >100 μM for KAT5, p300, pCAF and GCN5, respectively). Inhibits protein acetylation in prostate cancer cell lines and blocks DNA damage response. Decreases proliferation of LNCaP cells; induces apoptosis via caspase activation.
Spezifikation
Spezifikation
Chemischer Name oder Material | NU 9056 |
CAS | 1450644-28-6 |
Summenformel | C6H4N2S4 |
Menge | 10 mg |
Synonym | 5-1,2-thiazol-5-yldisulfanyl-1,2-thiazole, 1,2-bis isothiazol-5-yl disulfane |
InChI-Schlüssel | MLRAMCAHIWHWRM-UHFFFAOYSA-N |
Molekulargewicht (g/mol) | 232.35 |
PubChem CID | 72201043 |
Formelmasse | 232.37 |
Reinheit (%) | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'