Learn More
N-Benzoyl-2'-Desoxycytidin, ≥ 98 %, Thermo Scientific Chemicals
Beschreibung
Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Chemischer Name oder Material | N-Benzoyl-2′-deoxycytidine |
Verpackung | Glasflasche |
Menge | 250 mg |
Löslichkeitsinformationen | Soluble in methanol (50mg/ml). |
Formelmasse | 331.33 |
Reinheit (%) | ≥98% |
Physikalische Form | Fest |
RUO – Nur für Forschungszwecke
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.