missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fibronectin, ≥90%
Derived from Bovine Plasma
Marke: MP Biomedicals™ 0215002501
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Adhesive glycoproteins and are found both as cell surface proteins and in plasma. They are large glycoproteins found in cells and extracellular fluids. They are involved in a wide range of biological activities including cell adhesion to extracellular matrices, blood coagulation and wound healing
- Format: White lyophilized powder
- Molecular weight: 220,000 (Lit.)
- Soluble in water
Dentistry; Cell Biology and Analysis
Spezifikation
0.5 to 50μg/mL | |
220000 | |
1mg | |
4°C |
Cell Biology and Analysis | |
Fibronectin from Bovine Plasma | |
Bovine Plasma | |
Lyophilized |