Learn More
MP Biomedicals™ MP Biomedicals, Aprotinin , From Bovine Lung
From Bovine Lung
Marke: MP Biomedicals™ 0219038210
Beschreibung
Activity: Approx. 6000 Kallikrein-inhibitor-units per mg (KIU).
Unit Definition: One KIU (Kallikrein Inactivating Unit) is identical to the quantity of protease inhibitor that has the ability to inhibit 2 Kallikrein units by 50% under optimal conditions.
Aprotinin , [9087-70-1]; Pancreatic Basic Trypsin Inhibitor, polypeptide
- C284H432N84O79S7, F.W. 6500
- Analytical Grade
- Lyophilized white powder
Analytical

Spezifikation
1 to 2μg/mL | |
6512.42 | |
5 to 7 | |
Bovine Lung | |
Lyophilized |
Cell Biology and Analysis | |
Aprotinin from Bovine Lung | |
10 mg | |
4°C |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.