missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Metal Spare Tank
Marke: Carl Roth™ HX76.1
Weitere Details : Netto-Gewicht : 0.99000kg
Spezifikation
Spare Tank | |
Metal | |
For Consecutive 6-Digit Numbering, Possible to Print Without The Number Zero Infront | |
155 x 45 mm |
Black | |
With Self-Inking Pad | |
1 Pk. | |
7 g |