missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM5 Monoclonal Antibody (CRCT5.1 (A2.7A3)), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA191112
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.Spezifikation
MCM5 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P33992 | |
MCM5 | |
Bacterially expressed human Mcm5 | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
CRCT5.1 (A2.7A3) | |
Unconjugated | |
MCM5 | |
AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46 | |
Mouse | |
Protein G | |
RUO | |
4174 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |