Learn More
mCherry Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke:  Novus Biologicals NBP2-25157
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18025544
  • 601.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Knockdown Validated, Fluorescence Imaging.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
mCherry
Polyclonal
Unconjugated
50% PBS, 50% glycerol with 5mM Sodium Azide
Rabbit
27 kDa
0.1 mL
Cellular Markers, Fusion Proteins
This antibody detects mCherry.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
1 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated, Fluorescence Imaging
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
This mCherry Antibody was developed against full length recombinant mCherry protein
Affinity Purified
RUO
Primary
Non-species specific, Mouse, Rat, Bacteria