Learn More
Lonza™ BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures
Used as a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold. BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures are manufactured in accordance with cGMP regulations.
Spezifikation
Inhalt und Lagerung -10° to -20°C
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
  • Suggested working concentration of Penicillin-Streptomycin is 100 units of Potassium Penicillin and 100 μg of Streptomycin Sulfate per 1 ml of culture media
  • Manufactured in accordance with cGMP regulations
  • Cell culture tested to assure compatibility.
  • 20mL vials have crimp-top closure
  • 4.5, 100 and 500mL bottles have screw-cap closure
Spezifikation
Spezifikation
-10° to -20°C
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen