Learn More
Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnaces
Utilize the latest technical advances in heating elements, insulation and temperature control integrated into a self-contained cabinet with these box furnaces.
13266.00 €
Spezifikation
Temperatur (metrisch) 100 °C bis 1200 °C
Elektrische Anforderungen 208/240 V 50/60 Hz
Spannung 208/240 V
Frequenz 50/60 Hz
Mehr Spezifikationen ansehen
Produkte 1
Produktcode Marke Kapazität (metrisch) Temperaturregelung Preis Menge & Verfügbarkeit  
Produktcode Marke Kapazität (metrisch) Temperaturregelung Preis Menge & Verfügbarkeit  
17829393
Unterlagen ansehen Aktionsdetails
Thermo Scientific™
BF51842PBC-1
55.3 L Digital Multi Seg/Multi Prog/OTC
13266.00 €
1 Stück
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Utilize the latest technical advances in heating elements, insulation and temperature control integrated into a self-contained cabinet with Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnaces. Furnaces feature proprietary LGO (light gauge overbend) heating elements and Thermo Scientific™ Moldatherm™ insulation for efficient and economical transfer of heat to chamber, with low exterior temperatures. Variable heat-up rate eliminates thermal shock to materials with inherently quick heat up and cool down rates.
Key Features
  • Choice of side hinge or vertical lift door
  • Air vent (1 in. dia., top) and air inlet (0.375 in. dia., rear) for inert atmosphere exchange (may experience some leakage at door)
  • Self-tuning, digital instrumentation for precise temperature setpoint and display
  • Platinel II™ thermocouple for long-term stability
  • 0.6 cu. ft. models feature vertical lift door; 2 cu. ft. models feature horizontal side swing door, hot side facing away from operator for protection
Microprocessor Control
  • Microprocessor-based self-tuning PID control (proportional, integral, derivative) provides optimum thermal process, prevents overshoot
  • Control panel designed for easy access and maintenance
  • Main power ON/OFF switch on control panel
  • Adjustable high-limit overtemperature protection
  • Simultaneous LED display of actual temperature vs. setpoint
  • Can be configured to display temperature in either °C or °F
  • Safety door switch interrupts power to heating element when door is opened; protects heating elements and minimizes exposure to end-user
  • Removable shelves for versatility
  • Moldatherm™ hearthplate supports load and prevents damage due to spillage
Digital, Single-Setpoint Controller
  • Single segment, single setpoint, one ramp to setpoint
Digital Single-Program, Multiple-Segment Programmable Controller
  • Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
Digital Multiple-Program, Multiple-Segment Programmable Controller
Available on models with (P) suffix designation
  • Multiple programs and segments for ramp (up and down) and dwell (timed hold) temperature control
Overtemperature Control (OTC)†
Adjustable digital overtemperature control, available on selected models with(B)suffix designation
  • Protects furnace and load in the event of primary control circuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Manual reset required for safety
  • Operates via magnetic contacts through signal from independent thermocouple
Flowmeter Option (FM) (Inert Atmosphere Only)
Available on selected models with (FM) designation
  • Gas flowmeter, adjustable, located on front control panel
  • Adjustable flowrate, range 1.0 to 10.0 cu. ft./hr. standard
  • Suitable for inert gas or airflow to chamber
  • Allows fresh air exchange for ashing applications Not suitable for combustible or volatile gases
Ordering Information:
Required power cord and hardwiring not included. Optional RS485 Digital Communications Port allows controller to be connected to a PC for remote monitoring and control of the furnace; up to 30 units can be connected to one PC.
†Specify Option
B for Overtemperature Control (OTC) when ordering.
Includes
One two-part shelf (0.6 cu. ft. models have one shelf position at center position; 2.0 cu. ft. models have three shelf positions)
Spezifikation
Spezifikation
100 °C bis 1200 °C
208/240 V 50/60 Hz
208/240 V
50/60 Hz
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen