missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr5 Rat anti-Human, Oligo, Clone: 4D11F8 (also known as 4D11), BD™ AbSeq ™
Rat Monoclonal Antibody
Marke: BD Biosciences 940388
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Lgr5 Monoclonal antibody specifically detects Lgr5 in Human samples. It is validated for Single Cell 3' SequencingSpezifikation
Lgr5 | |
Monoclonal | |
Oligo | |
LGR5; FEX; HG38; GPR49; GPR67; GRP49 | |
Human LGR5 DNA | |
RUO | |
Human | |
IgG2b λ |
Single Cell 3' Sequencing | |
Clone 4D11F8 (also known as 4D11) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |