Learn More
LDHB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527505
Beschreibung
Recommended positive controls: 293T, U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (81%), Dog (100%), Pig (96%), Rabbit (100%), Chicken (87%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Spezifikation
LDHB | |
Polyclonal | |
Unconjugated | |
LDHB | |
AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16832, 24534, 3945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P07195, P16125, P42123 | |
LDHB | |
Recombinant fragment corresponding to a region within amino acids 118 and 334 of Human LDH-B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.