missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lamin A + C Antibody Blocking Peptide

Artikelnummer. 18535762 Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
0,1 mg
Packungsgröße:
0.10 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18535762

Marke: Novus Biologicals™ NB10056649PEP

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Specifications

Gene ID (Entrez) 4000
Konzentration 1 mg/mL
Inhalt und Lagerung Store at -80°C. Avoid freeze/thaw cycles.
Zur Verwendung mit (Anwendung) This Peptide is Useful as a Blocking Peptide for NB100-56649
Gen-Alias CDCD1, CDDC, CMD1A, CMT2B1, dilated 1 A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1 B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Gensymbol LMNA
Menge 0,1 mg
Forschungskategorie Apoptosis, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Protein Phosphatase, Signal Transduction, Stem Cells
Lagerungspuffer Peptide dissolved in dH20. Contains no BSA.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.