missing translation for 'onlineSavingsMsg'
Learn More

Insulin Receptor (β-subunit) Ab-5 Mouse Monoclonal Antibody, Epredia™

Epredia™ Insulin Receptor (β-Subunit) Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marke:  Epredia MS-635-P1

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 12611857

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: CT-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat.

Epitope: aa-GRVLTLPRS- within the C-terminus

Immunogen: A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor β-subunit

Molecular Weight: 95kDa

Positive Control: IM-9 lymphocyte cells

Cellular Localization: Membrane Ab-5 reacts with autophosphorylated and unoccupied receptors as well as the receptor-insulin complex. Ab-5 does not affect insulin-stimulated autophosphorylation.

Recommended for:

  • Immunoprecipitation
  • Affinity Purification (With peptide elution)
  • ELISA (As capture antibody only)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

Insulin Receptor (beta-Subunit) Ab-5
Monoclonal
200μg/mL
Mouse
500 μL
Steroids Hormones and Growth Factors
Human, Mouse, Rat
IgG2a κ
Immunoprecipitation
CT-1
Unconjugated
A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor Beta-subunit
RUO
Primary
Concentrated
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote