Learn More
Heat Shock Protein 70 (HSP70) Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.
133.62 € - 735.00 €
Spezifikation
Antigen | Heat Shock Protein 70 (HSP70) Ab-2 |
---|---|
Klon | W27 |
Anwendungen | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Klassifikation | Monoclonal |
Wirtsspezies | Mouse |
Produktcode | Marke | Form | Konjugat | Menge | Preis | Menge & Verfügbarkeit | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Form | Konjugat | Menge | Preis | Menge & Verfügbarkeit | |||
12621547
|
Epredia
MS-482-B0 |
Concentrated | Biotin | 100 μL | N/A | ||||
12631547
|
Epredia
MS-482-B1 |
Concentrated | Biotin | 500 μL | N/A | ||||
12651547
|
Epredia
MS-482-P0 |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12681547
|
Epredia
MS-482-PABX |
Concentrated | Unconjugated | 200 μL | N/A | ||||
12661547
|
Epredia
MS-482-P1 |
Concentrated | Unconjugated | 500 μL | N/A | ||||
12641547
|
Epredia
MS-482-P |
Concentrated | Unconjugated | 1 mL |
677.00 €
1 Milliliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||
12621557
|
Epredia
MS-482-R7 |
Prediluted | Unconjugated | 7 mL | N/A | ||||
Beschreibung
Heat shock proteins (HSP's) or stress response proteins (SRP's) are synthesized in variety of environmental and pathophysiological stressful conditions. Many HSP's are involved in processes such as protein denaturation-renaturation, folding-unfolding, transport-translocation, activation-inactivation, and secretion. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides, and other unknown proteins. Also, HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs, and other damaging conditions.Host Species: Mouse
Clone: W27
Isotype: IgG2a
Species Reactivity: Human, Monkey and Cow. Others not known.
Epitope: Not determined
Immunogen: HSP70 protein from HeLa cells
Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)
Positive Control: BT474 or HeLa cells, Breast CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Spezifikation
Heat Shock Protein 70 (HSP70) Ab-2 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Mouse | |
Stress Response | |
HSP70 protein from HeLa cells | |
Primary |
W27 | |
Monoclonal | |
RUO | |
Bovine, Human, Non-human Primate | |
IgG2a |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.