Learn More
DiscoverX KILR™ HCT-116 Cell Pool
Artikelnummer. 17314879
Encompass
Click to view available options
Menge:
2 Fläschchen
Artikelnummer. 17314879
Marke: DiscoverX 971040P047
om dit product te kopen Registreer vandaag om een webaccount aan te maken
The KILR™ HCT-116 Cell Pool allows for assay of effector-induced cell death by a variety of immune mechanisms. This cell pool ships as two vials of cryopreserved cells.
The KILR HCT-116 cell pool is a stable, engineered target cell pool which more closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint inhibitors, checkpoint agonists, bispecifics, CAR-T, and more) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of HCT-116 cells that overexpress a PL-tagged housekeeping gene. Once the cells have been lysed by effector cells, PL is released to the media, where EA contained in the detection kit produces an EFC signal proportional to the level of PL released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells.
Spezifikation
Assay-Eingabe Cytoxicity
Zelllinie HCT-116
Zur Verwendung mit (Anwendung) Cell Pools
Menge 2 Fläschchen