Learn More
BINDER™ Interior Lighting
Interior lighting with climate-resistant LEDs.
Marke: BINDER™ 8012-2565
Beschreibung
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Spezifikation
KBF 720, KBF 1060 | |
Glass | |
Lighting |