Learn More
Elma™ Insert Covers
Click to view available options
Modell:
100/120
150/180/300
180
30/40
60
80
Packungsgröße:
1 Stück
Beschreibung
- Elma ultrasonic units can be operated with or without a cover
- For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
- Covers make it possible to clean objects made of different materials simultaneously – but separately
- If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
- Allows cleaning with aggressive fluids
- Prolongs the service life of the unit
Spezifikation
Spezifikation
| Typ | Lid |
| Zur Verwendung mit (Geräte) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic S, Elmasonic Med, Elmasonic Select, Elmasonic EASY |
| Modell | 100/120 |