Learn More
Proteintech™ HumanKine™ Recombinant Human HGF Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Marke:  Proteintech™ HZ-1084-1000UG
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16811306
  • 2087.00 € / 1000 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spezifikation
Spezifikation
3082
1000 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 20 ng/mL EC50
Lyophilized
70 kDa, single chain, glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%