missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TSG Recombinant Protein, PeproTech®
Click to view available options
Menge:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
Recombinant Human TSG is a 198 amino acid, 22.2 kDa protein containing the BMP/TGF-beta binding portion of the full length TSG protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.
Spezifikation
Spezifikation
| Zugriffsnummer | Q9GZX9 |
| Zur Verwendung mit (Anwendung) | Functional Assay |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 57045 |
| Molekulargewicht | 22.2 kDa |
| Name | Human TSG |
| Menge | 50 μg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur