Learn More
STEMCELL Technologies Human Recombinant SCF
Artikelnummer. 17129481
Encompass_Preferred
Click to view available options
Menge:
10 μg
100 μg
1000 μg
Packungsgröße:
10 Mikrogramm
100 Mikrogramm
Artikelnummer. 17129481
Marke: STEMCELL Technologies 78062.2
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Stem cell factor
  • Stem cell factor (SCF) is an early-acting cytokine that plays a pivotal role in the regulation of embryonic and adult hematopoiesis
  • SCF promotes cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy
  • Together with other cytokines such as thrombopoietin and Flt3/Flk-2 Ligand, SCF is commonly used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture
  • In synergy with various growth factors, including IL-2, IL-3, IL-6, IL-7, G-CSF, and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells
  • SCF is also a primary growth and activation factor for mast cells and eosinophils
Spezifikation
Name Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor
Menge 1000 μg
Zelltyp Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Reinheits- oder Qualitätsgrad ≥95%