missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDX-1/IPF1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 27 publications
Marke: R&D Systems AF2419
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PDX-1/IPF1 Polyclonal specifically detects PDX-1/IPF1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Spezifikation
PDX-1/IPF1 | |
Polyclonal | |
Unconjugated | |
P52945 | |
PDX1 | |
E. coli-derived recombinant human PDX-1/IPF1 Ala91-Arg283 Accession # P52945 | |
100 μg | |
Primary | |
Detects human PDX-1/IPF1 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse PDX-1 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Glucose-sensitive factor, IDX1, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
Goat | |
Affinity Purified | |
RUO | |
3651 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur