missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Tenascin C Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 11 publications
Marke: R&D Systems MAB2138
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Tenascin C Monoclonal specifically detects Tenascin C in Human, Mouse samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.Spezifikation
Tenascin C | |
Monoclonal | |
LYOPH | |
Western Blot, Neutralization 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXB, HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin J1, Tenascin-C, tenascin-C isoform 14/AD1/16, TNC, TN-C, TNGP | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3371 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Neutralization, Immunocytochemistry | |
578 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TNC | |
Mouse immature astrocyte-derived Tenascin C | |
100 μg | |
Primary | |
Detect human and mouse Tenascin C in Western blots. | |
Human, Mouse | |
Purified |