missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LTF/Lactoferrin ELISA Kit

ELISA

692.00 € - 2780.00 €

Spezifikation

Zugriffsnummer Rinderlactoferrin; CKRX; Csp82; Epididymis luminales Protein 110; GIG12; wachstumshemmendes Protein 12; HEL 110; HLF2; Kaliocin 1; Kaliocin1; Kaliocin-1; Lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; Lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; Lfcin H; Lfcin-H; LTF; Minisatellit 10 r durch Sonde MMS10 erkannt; MMS10R; Ms10r; N-Lappen; neutrophiles Lactoferrin; Talalactoferrin
Assay-Bereich 1,96 bis 480 ng/ml
Assay-Empfindlichkeit: 1,96 ng/mL
Konjugat Biotin
Produkttyp ELISA
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17155079
Unterlagen ansehen
Invitrogen™
EH309RBX5
5 x 96 Tests
2780.00 €
1 Stück
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
16672103
Unterlagen ansehen
Invitrogen™
EH309RB
96 Tests
692.00 €
96 Tests
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung

Beschreibung

Human LTF/Lactoferrin quantitates human LTF/Lactoferrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LTF/Lactoferrin.

Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
Spezifikation

Spezifikation

Rinderlactoferrin; CKRX; Csp82; Epididymis luminales Protein 110; GIG12; wachstumshemmendes Protein 12; HEL 110; HLF2; Kaliocin 1; Kaliocin1; Kaliocin-1; Lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; Lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; Lfcin H; Lfcin-H; LTF; Minisatellit 10 r durch Sonde MMS10 erkannt; MMS10R; Ms10r; N-Lappen; neutrophiles Lactoferrin; Talalactoferrin
1,96 ng/mL
ELISA
Kolorimetrisches Mikrotiterplatten-Lesegerät
LTF
<10%
HRP
Überstand, 50 μl; Serum, 50 μl; Plasma, 50 μl
Lactoferrin
4 h 45 min
1,96 bis 480 ng/ml
Biotin
Plasma, Serum, Überstand
P02788.
<12%
Vorbeschichtete 96-Mikrotiterplatte, Standard, Assay-Verdünnungskonzentrat, biotinylierter Nachweisantikörper, SAV-HRP, Waschpuffer, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Nur für Forschungszwecke
2 bis 8 °C
1 h 20 min
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt