missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EGF (Q6QBS2, 53 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Marke: Abnova™ P3960.500ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq]
Spezifikation
Q6QBS2 | |
Lyophilized | |
6.2kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
EGF | |
The ED50 was determined by the dose-dependent stimulation of the proliferation of thymidine uptake by BALB/c 3T3 cells is < 0.1ng/mL, corresponding to a specific activity of > 1 x 107 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
1950 | |
EGF (Human) Recombinant Protein | |
500 μg | |
RUO | |
HOMG4/URG | |
EGF | |
E. coli | |
None | |
Lyophilized |