missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human DKK-1 Recombinant Protein, PeproTech®
Click to view available options
Menge:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
Mature human DKK-1 expressed in HEK293 cells is a 35-40 kDa glycoprotein containing 235 amino acid residues. The calculated molecular weight of Recombinant Human DKK-1 expressed in HEK293 cells is 25.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
Spezifikation
Spezifikation
| Zugriffsnummer | O94907 |
| Zur Verwendung mit (Anwendung) | Functional Assay |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 22943 |
| Molekulargewicht | 25.8 kDa |
| Name | Human DKK-1 |
| Menge | 50 μg |
| Quelle | HEK293 cells |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur