missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-1/CD80 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
242.00 € - 2140.00 €
Spezifikation
| Antigen | B7-1/CD80 |
|---|---|
| Klon | 37711 |
| Konzentration | LYOPH |
| Verdünnung | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.2-1 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF, In Situ Hybridization (ISH), ELISA |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18792769
![]() |
R&D Systems
MAB140-SP |
25 μg |
242.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18772709
![]() |
R&D Systems
MAB140-100 |
100 μg |
534.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18735809
![]() |
R&D Systems
MAB140-500 |
500 μg |
2140.00 €
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready, Dual RNAscope ISH-IHC, ELISA Capture (Matched Antibody Pair).Spezifikation
| B7-1/CD80 | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF, In Situ Hybridization (ISH), ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| CD80 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 37711 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.2-1 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P33681 | |
| 941 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-1/CD80 Val35-Asn242 (predicted) Accession # P33681 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human B7-1/CD80 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) B7-2, recombinant mouse (rm) B7-1, rmB7-2, rhB7-H1, rhB7-H2, rhB7-H3 or rmPD-L2 is observed. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts