Learn More
Invitrogen™ HLA-BC Monoclonal Antibody (B1.23.2), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 16-5935-38
Beskrivelse
Description: This B1.23.2 monoclonal antibody recognizes the human Human Leukocyte Antigens (HLA) B and C. Along with HLA-A, these proteins correspond to MHC Class I in humans. HLA-B and HLA-C are expressed on nearly all cell types and heterodimerize with beta-2 microglobulin at the cell surface. These molecular complexes play a critical role in the presentation of endoplasmic reticulum-derived peptides to immune cells. The antibody has been shown to inhibit binding of KIR2DL1 and LIR1. Applications Reported: This B1.23.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B1.23.2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
HLA-class I antigens are widely distributed on human nucleated cells. The intensity of HLA-ABC maybe altered in pathological states e.g. malignant cells may lose HLA-ABC, whereas hepatocytes in alcoholic hepatitis, biliary cirrhosis and chronic active hepatitis may show enhanced expression of HLA-ABC. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Tekniske data
| HLA-BC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P01889 | |
| HLA-B | |
| Affinity Chromatography | |
| RUO | |
| 3106 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Functional Assay | |
| B1.23.2 | |
| Functional Grade | |
| HLA-B | |
| AS; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; epididymis secretory protein Li 83; HEL-S-83; histocompatibility antigen; HLA -B; HLA B-40011; HLA B44; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAB; HLA-B; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLA-Cw; HLA-DRB1; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility complex, class I, B; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; SPDA1; truncated class I antigen; truncated MHC class I antigen | |
| Mouse | |
| 5 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.