missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-2 alpha/EPAS1 Antibody, PE/Cy7, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB100-122PECY7
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat, Fish, Hamster, Primate, Rabbit, Reptile, Sheep samples. It is validated for Flow Cytometry, Knockout Validated, Knockdown Validated.
Spezifikation
| HIF-2 alpha/EPAS1 | |
| Polyclonal | |
| Flow Cytometry, Knockout Validated, Knockdown Validated | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, | |
| Rabbit | |
| Affinity Purified | |
| Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 2034.0 | |
| Human, Mouse, Rat, Fish, Hamster, Primate, Rabbit, Reptile, Sheep | |
| IgG |
| Flow Cytometry, Immunoassay, KnockDown | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| EPAS1 | |
| This HIF-2 alpha/EPAS1 Antibody was developed against a peptide derived from the C-terminus of mouse/human HIF-2 alpha protein. | |
| 0.1 mL | |
| Primary | |
| This HIF-2 alpha/EPAS1 Antibody is specific for HIF-2 alpha/EPAS, and does not cross-react with HIF-1 alpha. | |
| Store at 4°C in the dark. Do not freeze. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur