missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16842-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Product No. 16842-1-AP is a rabbit polyclonal antibody raised against the full-length of human HEBP1. The MW of this protein is 21 kDa, and this antibody specifically recognizes the 21 kDa protein.
The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells.Spezifikation
HEBP1 | |
Polyclonal | |
Unconjugated | |
HEBP1 | |
HBP, HEBP, HEBP1, heme binding protein 1, p22HBP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15199, 362454, 50865 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NRV9, Q9R257 | |
Hebp1 | |
HEBP1 Fusion Protein Ag10538 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur