missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Tocris Bioscience™
Click to view available options
Menge:
1 g
Beschreibung
One of the major inhibitory neurotransmitters in the mammalian CNS, predominantly active in the spinal cord and brain stem. Also acts as a modulator of excitatory amino acid transmission mediated by NMDA receptors. Also available as part of the NMDA Receptor - Glycine Site Tocriset™ .
Spezifikation
Spezifikation
Chemischer Name oder Material | Glycine |
CAS | 56-40-6 |
Summenformel | C2H5NO2 |
MDL-Nummer | MFCD00008131 |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
InChI-Schlüssel | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
SMILES | NCC(O)=O |
IUPAC-Name | 2-Aminoessigsäure |
Molekulargewicht (g/mol) | 75.07 |
PubChem CID | 750 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'