Learn More
Kova International™ Glasstic Slide with Counting Grids
A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic with a Neubauer counting grid.
Marke:  Kova International™ 87144E
Weitere Details : Netto-Gewicht : 0.43200kg
Artikelnummer. 10298483
  • 129.85 € / 1 Stück
Verfügbar ab: 19-08-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • The slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
  • Reduces pipetting errors and eliminates variables that can interfere with microscopic urinalysis
  • Superior optical plastic with a refractive index that is similar to glass
  • Manufactured to the specifications of high quality optical lenses, smooth with a tolerance below 50
  • Perimeter contained wells for precise volume control and protection against cross-contamination and leakage
  • Easier and faster counting with Neubauer-style ruling pattern
  • Capillary action provides reproducible precision
  • Homogeneous suspension of sample sediments
Spezifikation
Spezifikation
Slide with Counting Grid
10
Transparent
Microscopy applications
Optical Plastic
Not Frosted
Neubauer style ruling pattern
100