missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAL4 Monoclonal Antibody

Mouse Monoclonal Antibody
Marke: Invitrogen™ 338600
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
33-8600 detects the Gal4 DNA Binding Domain by western blot, immunoprecipitation and immunofluorescence corresponding to amino acids 1-147 of the GAL4 DNA binding region.
GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a 'zinc finger' sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.
Spezifikation
| GAL4 | |
| Monoclonal | |
| Unconjugated | |
| GAL4 | |
| antigen NY-CO-27; GAL4; Gal-4; GAL81; galactose-responsive transcription factor GAL4; galectin 4; galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; LGALS4; Regulatory protein GAL4; YPL248C | |
| Mouse | |
| Protein A | |
| RUO | |
| 855828 | |
| -20°C | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P04386 | |
| GAL4 | |
| A recombinant protein corresponding to amino acids 1-147 mapping within the amino terminal, DNA binding domain of GAL4. | |
| 200 μg | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur