missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorid-Standardlösung (Ion HIQU), Chem Lab™
Click to view available options
Menge:
500 ml
Spezifikation
Spezifikation
Menge | 500 ml |
Farbe | Farblos |
Geruch | Geruchsneutral |
pH | >11 |
Löslichkeit | Wasser |
Physikalische Form | Flüssig |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'