Learn More
Fisherbrand™ SafeSnap™ Disposable Ampule Openers
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Spezifikation
Einwegprodukt | Ja |
---|---|
Produkttyp | Ampullenöffner |
Material | Polyethylen |
Beschreibung
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
- To use: Slip the collar onto the ampule until collar rests on the widest part
- Grip the collar with thumb and forefinger while holding the upright ampule at bottom
- Break the ampule with a quick snap
- Dispose of the collar and ampule top together
Spezifikation
Ja | |
Ampullenöffner | |
Polyethylen |