Learn More
Abnova™ FGF7 (Human) Recombinant Protein
Human FGF7 (180 a.a.) full-lenth recombinant protein with His tag expressed in Barley grain (Hordeum vulgare). This protein is endotoxins-free and thus optimal to use in stem cell cultures.
Marke: Abnova™ P5880.50ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
2252 | |
Hordeum vulgare (barley) expression system | |
50 μg | |
HBGF-7, KGF | |
Human | |
His | |
Lyophilized |
fibroblast growth factor 7 (keratinocyte growth factor) | |
SDS-PAGE Stained with Coomassie Blue | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
FGF7 | |
Plants | |
Lyophilized from PBS, pH 7.2 |