Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, polypropylene 13ga x 90mm, sterile
Artikelnummer. 15145014
Encompass
Click to view available options
Produkttyp:
Zulaufschlauch
Artikelnummer. 15145014
Marke: Instech Laboratories™ FTP139050
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Instech's polypropylene gavage tubes are flexible to reduce trauma, translucent to visualise delivery, and disposable to eliminate the cost of cleaning and the chance of compound crossover. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes. The tube and luer hub are made of the same material used to make plastic syringes. The soft, bulb-shaped tip is designed to reduce the chances of inadvertent placement in the trachea and perforation of the oesophagus. Available in 11 sizes for rodents ranging from mouse pups to rats to rabbits.
  • Plastic feeding tubes, 13ga x 90mm
  • For rodent oral gavage
  • Designed to minimise trauma to oesophagus
  • Polypropylene female luer (purple) and flexible polypropylene tube with soft elastomer tip
  • Disposable
  • Supplied as 10 sterile pouches of 5 tubes
Spezifikation
Produkttyp Zulaufschlauch