missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterovirus 71 3AB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP3-13091
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Enterovirus 71 3AB Polyclonal antibody specifically detects Enterovirus 71 3AB in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spezifikation
| Enterovirus 71 3AB | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| EV71 3AB, Protein 3AB | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Enterovirus 71 3AB (EV71 virus (strain USA/BrCr/1970)). The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction