Learn More
STEMCELL Technologies EasySep™ Human PE Positive Selection Kit II
Click to view available options
Menge:
1 x 109 Zellen
5 x 109 Zellen
Unit Size:
1 Set
Description
- The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection
- Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Human PE Positive Selection Kit (Catalog #18551)
Specifications
Specifications
| Zelltyp | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 1 x 109 Zellen |