missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 51005-2-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. DUSP22 dephosphorylates ERK2 MAP kinase and JNK. DUSP22 displays highest in thymus, but it is also detectable in monocytes and lymphocytes.Spezifikation
DUSP22 | |
Polyclonal | |
Unconjugated | |
DUSP22 | |
DUSP22, JKAP, JNK stimulatory phosphatase 1, JSP 1, JSP1, LMW DSP2, LMWDSP2, MAP kinase phosphatase x, MKP x, MKPX, VHX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56940 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRW4 | |
DUSP22 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |