missing translation for 'onlineSavingsMsg'
Learn More

DUSP22 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 51005-2-AP-150UL

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16876255

  • 410.97 € / 150 Mikroliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. DUSP22 dephosphorylates ERK2 MAP kinase and JNK. DUSP22 displays highest in thymus, but it is also detectable in monocytes and lymphocytes.
Spezifikation

Spezifikation

DUSP22
Polyclonal
Unconjugated
DUSP22
DUSP22, JKAP, JNK stimulatory phosphatase 1, JSP 1, JSP1, LMW DSP2, LMWDSP2, MAP kinase phosphatase x, MKP x, MKPX, VHX
Rabbit
Antigen Affinity Chromatography
RUO
56940
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NRW4
DUSP22
Peptide
150 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote