missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline hyclate, Tocris Bioscience™
Click to view available options
Menge:
50 mg
Beschreibung
Broad-spectrum antibiotic and MMP inhibitor; tetracycline derivative. Increases and promotes smooth muscle cell adhesion; inhibits proliferation and migration. Protects the microvasculature by inhibiting plasminogen systems.
Spezifikation
Spezifikation
Chemischer Name oder Material | Doxycycline hyclate |
CAS | 1293992-19-4 |
Summenformel | C22H24N2O8·HCl |
Menge | 50 mg |
Synonym | (4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)- 3,5,10,12,12a-pentahydroxy- 6-methyl- 1,11-dioxo- 1,4,4a,5,5a,6,11,12a-octahydrotetracene- 2-carboxamide hydrochloride |
Formelmasse | 480.9 |
Reinheit (%) | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'