missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DO-11.10 Clonotypic TCR Mouse anti-Mouse, Oligo, Clone: KJ1-26, BD™ AbSeq ™
Mouse Monoclonal Antibody
Marke: BD Biosciences 940479
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
DO-11.10 Clonotypic TCR Monoclonal antibody specifically detects DO-11.10 Clonotypic TCR in Mouse samples. It is validated for Single Cell 3' SequencingSpezifikation
DO-11.10 Clonotypic TCR | |
Monoclonal | |
Oligo | |
DO-11.10 clonotypic TCR | |
DO-11.10 T hybridoma cells | |
RUO | |
Mouse | |
IgG2a κ |
Single Cell 3' Sequencing | |
Clone KJ1-26 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |