Learn More
DNA Polymerase gamma Antibody (S06-1G0), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-19996-100ul
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DNA Polymerase gamma Monoclonal antibody specifically detects DNA Polymerase gamma in Human samples. It is validated for Western Blot
Spezifikation
| DNA Polymerase gamma | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S06-1G0 | |
| Unconjugated | |
| DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE | |
| A synthetic peptide of human DNA Polymerase gamma (Uniprot # P54098) | |
| 100 μg | |
| DNA Polymerases, DNA Repair | |
| 5428 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |