missing translation for 'onlineSavingsMsg'
Learn More

DL-TBOA, Tocris Bioscience™

Artikelnummer. 30001210
Click to view available options
Menge:
10 mg
50 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Selective non-transportable inhibitor of EAATs

DL-TBOA is a competitive, non-transportable blocker of excitatory amino acid transporters (IC50 values are 6 μM, 6 μM and 70 μM, for EAAT2, EAAT3 and EAAT1 respectively). DL-TBOA also inhibits EAAT4 and EAAT5 (Ki values are 4.4 μM and 3.2 μM respectively). DL-TBOA displays high selectivity for EAATs over ionotropic and metabotropic glutamate receptors. DL-TBOA augments SN 38 (Cat. No. 2684) induced colorectal cancer cell death in drug-resistant cell lines. DL-TBOA enhances synaptic activity in the frog labyrinth.

Chemische Identifikatoren

CAS 205309-81-5
Summenformel C11H13NO5
Molekulargewicht (g/mol) 239.227
InChI-Schlüssel BYOBCYXURWDEDS-IUCAKERBSA-N
Synonym dl-tboa, chembl79140, dl-threo-beta-benzyloxyaspartate, 3s-3-benzyloxy-l-aspartic acid, 2s,3s-2-amino-3-benzyloxy succinic acid, tboa, 2s,3s-2-amino-3-benzyloxy-succinic acid, tocris-1223, d07gub, l-threo-benzyloxy aspartate
PubChem CID 5311218
IUPAC-Name (2S,3S)-2-amino-3-phenylmethoxybutanedioic acid
SMILES C1=CC=C(C=C1)COC(C(C(=O)O)N)C(=O)O

Spezifikation

Chemischer Name oder Material DL-TBOA
Formelmasse 239.23
Reinheit (%) >98%
Menge 10 mg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'