Learn More
Tocris Bioscience™ Cy5B-trisulfo, NHS
Restricted conformation cyanine dye with high quantum yield and long fluorescence lifetime for advanced microscopy applications
Marke:  Tocris Bioscience™
308.10 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Artikelnummer. 18064442
  • 376.00 € / 300 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
Key information: Cy5B-trisulfo, NHS is a far-red fluorescent dye; supplied with an NHS ester reactive group for the labeling of primary amines. Particularly useful for smFRET experiments to reduce blinking and bleaching artefacts and maintain correct mean FRET efficiencies. Application: FRET, smFRET, FLIM, TIRF, and super resolution microscopy (SRM) techniques, including dSTORM. Provides enhanced spatial resolution in dSTORM experiments. Does not provide efficient blinking in conventional high-thiol dSTORM buffers; instead, sodium borohydride (NaBH4) addition is required following which SmLM experiments can be performed in PBS. Properties and Photophysical Data: Excitation and emission maxima (λ) are 669 nm and 685 nm, respectively; extinction coefficient = 241,000 M-1cm-1; quantum yield = 0.4; fluorescence lifetime = 1.85 ns. We suggest making up a 10 mM stock solution of this product in anhydrous DMSO. For more information on how Cy5B-trisulfo, NHS may be used, see our protocol: Conjugation Protocol for Amine Reactive Dyes .
Spezifikation
Spezifikation
300 μg
Fluorescent Dyes
C47H52N4Na2O15S3