Learn More
R&D Systems™ Cultrex Antibiotic-free Mouse Laminin I, Pathclear
Supports the attachment of MG63 osteosarcoma cells at <20 μg/mL.
Marke: R&D Systems™ 3401-010-02
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sterility Testing:
- PathClear™ - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses.
- No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP sterility testing guidelines.
- Endotoxin concentration < 20 EU/mL by LAL assay.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
1 mg | |
LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha |