missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clonidine hydrochloride, Tocris Bioscience™
Click to view available options
Menge:
100 mg
Beschreibung
Prototypical I1 imidazoline receptor ligand. α2-adrenergic receptor agonist. Antihypertensive. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.
Spezifikation
Spezifikation
Chemischer Name oder Material | Clonidine hydrochloride |
CAS | 4205-91-8 |
Summenformel | C9H10Cl3N3 |
Menge | 100 mg |
Synonym | clonidine hydrochloride, clonidine hcl, catapres, 2-2,6-dichloroanilino-2-imidazoline hydrochloride, chlophazolin, dispaclonidin, normopresan, atensina, barclyd, capresin |
InChI-Schlüssel | ZNIFSRGNXRYGHF-UHFFFAOYSA-N |
SMILES | C1CN=C(N1)NC2=C(C=CC=C2Cl)Cl.Cl |
IUPAC-Name | N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride |
Molekulargewicht (g/mol) | 266.55 |
PubChem CID | 20179 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'